Assay Details
| Gene Symbol: Kif24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024241.2 | NP_077203.2 | 7 - 8 | 1485 | 90 | |
| GenBank mRNA | AK050683.1 | - | 7 - 8 | 1471 | 90 | |
| AK084967.1 | - | 4 - 5 | 1084 | 90 | ||
| AK135307.1 | - | 7 - 8 | 1486 | 90 | ||
| BC067395.1 | - | 7 - 8 | 1477 | 30460089 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif24
Genomic Map