Assay Details
| Gene Symbol: Kif24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024241.2 | NP_077203.2 | 8 - 9 | 1592 | 73 | |
| GenBank mRNA | AK050683.1 | - | 8 - 9 | 1578 | 73 | |
| AK084967.1 | - | 5 - 6 | 1191 | 73 | ||
| AK135307.1 | - | 8 - 9 | 1593 | 73 | ||
| BC067395.1 | - | 8 - 9 | 1584 | 30460089 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif24
Genomic Map