Assay Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133911.1 | NP_598672.1 | 12 - 13 | 1994 | 75 | |
| GenBank mRNA | AK028929.1 | - | 12 - 13 | 1933 | 75 | |
| AK131924.1 | - | 1 - 2 | 136 | 75 | ||
| BC019649.1 | - | 1 - 2 | 121 | 75 | ||
| BC020317.2 | - | 7 - 8 | 1127 | 75 | ||
| BC052391.1 | - | 12 - 13 | 1994 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgra2
Genomic Map