Assay Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133911.1 | NP_598672.1 | 18 - 19 | 2908 | 97 | |
| GenBank mRNA | AK028929.1 | - | 18 - 19 | 2847 | 97 | |
| AK131924.1 | - | 7 - 8 | 1050 | 97 | ||
| AK195901.1 | - | 1 - 2 | 45 | 97 | ||
| AK197812.1 | - | 1 - 2 | 45 | 97 | ||
| AK210870.1 | - | 1 - 2 | 43 | 97 | ||
| BC019649.1 | - | 7 - 8 | 1035 | 97 | ||
| BC020317.2 | - | 13 - 14 | 2041 | 97 | ||
| BC048928.1 | - | 2 - 3 | 178 | 97 | ||
| BC052391.1 | - | 18 - 19 | 2908 | 97 | ||
| BC058251.1 | - | 2 - 3 | 178 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2173832 | BC052391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2173832 | BC020317.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2173832 | NM_133911.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2173832 | AK028929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377797 | XM_001473102.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377797 | XM_001474475.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4552 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | AK030000.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | AK131111.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | BC025196.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | BC032200.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | BC051105.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | BC089363.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375444 | NM_146205.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108088 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542267 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11407 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141323 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158701 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137914 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2173832 | BC052391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2173832 | BC020317.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2173832 | NM_133911.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2173832 | AK028929.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01097076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137914 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158701 | - | - |
| Whole Mouse Genome G4122A | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC048928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC052391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC058251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC019649.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462102 | BC020317.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137914 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158701 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04360956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04123098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04541672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04360967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04754678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04754670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04692193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04360951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04626589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04754701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04262332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04754684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04360962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23680981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23681046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10529960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529967 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529966 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529974 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529975 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529968 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529970 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529965 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529958 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444300_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431034_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426782_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10529957 | - | - |
| Mouse Genome 430 2.0 Array | 1431034_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426782_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431034_at | - | - |
| Mouse Genome 430 2.0 Array | 1426782_at | - | - |
| Mouse Genome 430 2.0 Array | 1444300_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444300_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70693_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70693_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500021611.mm.1 | NM_133911.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021611.mm.1 | NM_133911.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021611.mm.1 | TC0500002386.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500021610.mm.1 | NM_133911.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021610.mm.1 | NM_133911.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500021610.mm.1 | TC0500002386.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431034_at | - | - |
| Mouse Expression Set 430 | 1431034_PM_at | - | - |
| Mouse Expression Set 430 | 1444300_at | - | - |
| Mouse Expression Set 430 | 1444300_PM_at | - | - |
| Murine Genome U74v2 Set | 103332_PM_at | - | - |
| Murine Genome U74v2 Set | 103332_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426782_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426782_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444300_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444300_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431034_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431034_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251856 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1251856 | - | - |

Adgra2
Genomic Map