Assay Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133911.1 | NP_598672.1 | 11 - 12 | 1797 | 81 | |
| GenBank mRNA | AK028929.1 | - | 11 - 12 | 1736 | 81 | |
| BC020317.2 | - | 6 - 7 | 930 | 81 | ||
| BC052391.1 | - | 11 - 12 | 1797 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgra2
Genomic Map