Assay Details
| Gene Symbol: Lipm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023903.1 | NP_076392.1 | 3 - 4 | 408 | 100 | |
| GenBank mRNA | AK019504.1 | - | 3 - 4 | 419 | 100 | |
| AK037091.1 | - | 3 - 4 | 410 | 100 | ||
| AK085719.1 | - | 3 - 4 | 384 | 100 | ||
| BC031933.1 | - | 3 - 4 | 378 | 4020396 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipm
Genomic Map