Assay Details
| Gene Symbol: Srp72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025691.1 | NP_079967.1 | 11 - 12 | 1182 | 86 | |
| GenBank mRNA | AK133784.1 | - | 11 - 12 | 1183 | 86 | |
| AK164641.1 | - | 5 - 6 | 540 | 86 | ||
| BC003434.1 | - | 11 - 12 | 1170 | 86 | ||
| BC019196.1 | - | 11 - 12 | 1175 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srp72
Genomic Map