Assay Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008066.3 | NP_032092.1 | 6 - 7 | 849 | 92 | |
| GenBank mRNA | AK039055.1 | - | 6 - 7 | 850 | 92 | |
| AK046953.1 | - | 1 - 2 | 983 | 92 | ||
| AK048165.1 | - | 747 | 92 | |||
| BC115727.1 | - | 6 - 7 | 771 | 40039253 | 92 | |
| M86567.1 | - | 6 - 7 | 600 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra2
Genomic Map