Assay Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177561.3 | NP_808229.1 | 3 - 4 | 447 | 77 | |
| GenBank mRNA | AK029604.1 | - | 2 - 3 | 299 | 77 | |
| AK145298.1 | - | 3 - 4 | 467 | 77 | ||
| BC030888.1 | - | 1 - 2 | 214 | 77 | ||
| BC039916.1 | - | 3 - 4 | 449 | 4950034 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp46
Genomic Map