Assay Details
| Gene Symbol: Aasdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173765.3 | NP_776126.1 | 12 - 13 | 2781 | 124 | |
| GenBank mRNA | AB095954.1 | - | 12 - 13 | 2798 | 124 | |
| AK136849.1 | - | 12 - 13 | 2781 | 124 | ||
| AK148405.1 | - | 11 - 12 | 2607 | 124 | ||
| BC058102.1 | - | 12 - 13 | 2766 | 124 | ||
| BC094507.1 | - | 10 - 11 | 3097 | 124 | ||
| BC128330.1 | - | 11 - 12 | 2653 | 40110405 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aasdh
Genomic Map