| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500023328.mm.1 |
NM_173765.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023328.mm.1 |
NM_173765.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023328.mm.1 |
TC0500002641.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500023335.mm.1 |
NM_173765.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023335.mm.1 |
NM_173765.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023335.mm.1 |
TC0500002641.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500023325.mm.1 |
NM_173765.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023325.mm.1 |
NM_173765.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023325.mm.1 |
TC0500002641.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500023327.mm.1 |
NM_173765.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023327.mm.1 |
NM_173765.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500023327.mm.1 |
TC0500002641.mm.1 |
MTA TC |