Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 1 - 2 | 133 | 76 | |
| GenBank mRNA | BC053023.1 | - | 1 - 2 | 87 | 6401682 | 76 |
| BC060104.1 | - | 1 - 2 | 93 | 76 | ||
| CJ064850.1 | - | 1 - 2 | 134 | 76 | ||
| L04302.1 | - | 1 - 2 | 113 | 76 | ||
| L24434.1 | - | 1 - 2 | 110 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thbs3
Genomic Map