Assay Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130905.2 | NP_570975.1 | 2 - 3 | 153 | 70 | |
| GenBank mRNA | AF373412.1 | - | 2 - 3 | 153 | 70 | |
| AK040917.1 | - | 2 - 3 | 175 | 70 | ||
| BC107188.2 | - | 2 - 3 | 153 | 40050810 | 70 | |
| BC107189.2 | - | 2 - 3 | 153 | 40050811 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd209e
Genomic Map