Assay Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010485.3 | NP_034615.2 | 1 - 2 | 215 | 86 | |
| GenBank mRNA | AK011870.1 | - | 1 - 2 | 168 | 86 | |
| AK034536.1 | - | 1 - 2 | 202 | 86 | ||
| AK075818.1 | - | 1 - 2 | 177 | 86 | ||
| AK080365.1 | - | 1 - 2 | 173 | 86 | ||
| AK148528.1 | - | 1 - 2 | 143 | 86 | ||
| BY775007.1 | - | 1 - 2 | 215 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034536.1, AK075818.1, AK080365.1, AK148528.1, NM_010485.3 transcripts.
Set Membership: |
|

Elavl1
Genomic Map