Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 15 - 16 | 2116 | 73 | |
| GenBank mRNA | AK002890.1 | - | 15 - 16 | 2117 | 73 | |
| AK031533.1 | - | 2203 | 73 | |||
| AK087952.1 | - | 12 - 13 | 2379 | 73 | ||
| AK132434.1 | - | 15 - 16 | 2130 | 73 | ||
| AK145925.1 | - | 15 - 16 | 2118 | 73 | ||
| BC021341.1 | - | 15 - 16 | 2114 | 5066146 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map