Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 9 - 10 | 1255 | 64 | |
| GenBank mRNA | AK002890.1 | - | 9 - 10 | 1256 | 64 | |
| AK031533.1 | - | 1265 | 64 | |||
| AK132434.1 | - | 9 - 10 | 1269 | 64 | ||
| AK145925.1 | - | 9 - 10 | 1257 | 64 | ||
| BC021341.1 | - | 9 - 10 | 1253 | 5066146 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map