Assay Details
| Gene Symbol: Ccl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009138.3 | NP_033164.1 | 1 - 2 | 66 | 69 | |
| GenBank mRNA | AJ249480.1 | - | 1 - 2 | 39 | 69 | |
| AK131875.1 | - | 1 - 2 | 67 | 69 | ||
| AK134138.1 | - | 1 - 2 | 51 | 69 | ||
| DQ158256.1 | - | 1 - 2 | 52 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ249480.1, AK131875.1, AK134138.1, DQ158256.1, NM_009138.3 transcripts.
Set Membership: |
|

Ccl25
Genomic Map