Assay Details
| Gene Symbol: Pet100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195244.1 | NP_001182173.1 | 1 - 2 | 59 | 100 | |
| GenBank mRNA | AK013674.1 | - | 1 - 2 | 61 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195244.1 transcripts.
Set Membership: |
|

Pet100
Genomic Map