Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 11 - 12 | 1254 | 105 | |
| NM_001286207.1 | NP_001273136.1 | 11 - 12 | 1227 | 105 | ||
| NM_011791.3 | NP_035921.2 | 11 - 12 | 1344 | 105 | ||
| GenBank mRNA | AK087934.1 | - | 11 - 12 | 1319 | 105 | |
| AK132471.1 | - | 10 - 11 | 1250 | 105 | ||
| AK145576.1 | - | 11 - 12 | 1219 | 105 | ||
| AK145790.1 | - | 11 - 12 | 1231 | 105 | ||
| AK145967.1 | - | 11 - 12 | 1198 | 105 | ||
| AK146599.1 | - | 11 - 12 | 1228 | 105 | ||
| AK146756.1 | - | 11 - 12 | 1219 | 105 | ||
| AK146938.1 | - | 11 - 12 | 1321 | 105 | ||
| BC012957.1 | - | 11 - 12 | 1331 | 4211433 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ash2l
Genomic Map