Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 4 - 5 | 410 | 71 | |
| NM_001286207.1 | NP_001273136.1 | 4 - 5 | 383 | 71 | ||
| NM_011791.3 | NP_035921.2 | 4 - 5 | 500 | 71 | ||
| GenBank mRNA | AB020983.2 | - | 4 - 5 | 482 | 71 | |
| AK087934.1 | - | 4 - 5 | 475 | 71 | ||
| AK132471.1 | - | 4 - 5 | 482 | 71 | ||
| AK145576.1 | - | 4 - 5 | 375 | 71 | ||
| AK145790.1 | - | 4 - 5 | 387 | 71 | ||
| AK145967.1 | - | 4 - 5 | 354 | 71 | ||
| AK146254.1 | - | 4 - 5 | 412 | 71 | ||
| AK146599.1 | - | 4 - 5 | 384 | 71 | ||
| AK146756.1 | - | 4 - 5 | 375 | 71 | ||
| AK146938.1 | - | 4 - 5 | 477 | 71 | ||
| BC012957.1 | - | 4 - 5 | 487 | 4211433 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ash2l
Genomic Map