Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 7 - 8 | 697 | 89 | |
| NM_001286207.1 | NP_001273136.1 | 7 - 8 | 670 | 89 | ||
| NM_011791.3 | NP_035921.2 | 7 - 8 | 787 | 89 | ||
| GenBank mRNA | AB020983.2 | - | 7 - 8 | 769 | 89 | |
| AK087934.1 | - | 7 - 8 | 762 | 89 | ||
| AK145576.1 | - | 7 - 8 | 662 | 89 | ||
| AK145790.1 | - | 7 - 8 | 674 | 89 | ||
| AK145967.1 | - | 7 - 8 | 641 | 89 | ||
| AK146254.1 | - | 7 - 8 | 699 | 89 | ||
| AK146599.1 | - | 7 - 8 | 671 | 89 | ||
| AK146756.1 | - | 7 - 8 | 662 | 89 | ||
| AK146938.1 | - | 7 - 8 | 764 | 89 | ||
| AK178767.1 | - | 3 - 4 | 243 | 89 | ||
| AK218371.1 | - | 3 - 4 | 243 | 89 | ||
| BC012957.1 | - | 7 - 8 | 774 | 4211433 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ash2l
Genomic Map