Assay Details
| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039466.1 | NP_001034555.1 | 6 - 7 | 1183 | 97 | |
| NM_183037.2 | NP_898858.1 | 6 - 7 | 1286 | 97 | ||
| NR_121683.1 | 6 - 7 | 1416 | 97 | |||
| GenBank mRNA | AK132368.1 | - | 6 - 7 | 1153 | 97 | |
| AK170904.1 | - | 6 - 7 | 1184 | 97 | ||
| AY251388.1 | - | 6 - 7 | 1166 | 97 | ||
| BC065049.1 | - | 6 - 7 | 1329 | 6418851 | 97 | |
| BC082299.1 | - | 6 - 7 | 1286 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim46
Genomic Map