Assay Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144898.2 | NP_659147.2 | 1 - 2 | 163 | 68 | |
| GenBank mRNA | AK139708.1 | - | 146 | 68 | ||
| AK166236.1 | - | 1 - 2 | 135 | 68 | ||
| AK167586.1 | - | 1 - 2 | 111 | 68 | ||
| AK170160.1 | - | 1 - 2 | 69 | |||
| BC008103.1 | - | 1 - 2 | 113 | 3966815 | 68 | |
| BC108354.1 | - | 1 - 2 | 97 | 5035560 | 68 | |
| BY267786.1 | - | 1 - 2 | 164 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msto1
Genomic Map