Assay Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144898.2 | NP_659147.2 | 5 - 6 | 501 | 113 | |
| GenBank mRNA | AK139708.1 | - | 480 | 113 | ||
| AK163280.1 | - | 5 - 6 | 361 | 113 | ||
| AK166236.1 | - | 5 - 6 | 473 | 113 | ||
| AK167586.1 | - | 5 - 6 | 449 | 113 | ||
| AK170160.1 | - | 5 - 6 | 484 | 113 | ||
| BC008103.1 | - | 5 - 6 | 451 | 3966815 | 113 | |
| BC108354.1 | - | 5 - 6 | 435 | 5035560 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msto1
Genomic Map