| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300005569.mm.1 |
NM_001014995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005569.mm.1 |
NM_001014995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005569.mm.1 |
TC0300000749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005573.mm.1 |
NM_001014995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005573.mm.1 |
NM_001014995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005573.mm.1 |
TC0300000749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005567.mm.1 |
NM_001014995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005567.mm.1 |
NM_001014995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005567.mm.1 |
TC0300000749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005568.mm.1 |
NM_001014995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005568.mm.1 |
NM_001014995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005568.mm.1 |
TC0300000749.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005571.mm.1 |
NM_001014995.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005571.mm.1 |
NM_001014995.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005571.mm.1 |
TC0300000749.mm.1 |
MTA TC |