Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163432.1 | NP_001156904.1 | 8 - 9 | 1226 | 101 | |
| NM_007712.3 | NP_031738.2 | 8 - 9 | 1229 | 101 | ||
| GenBank mRNA | AF033564.1 | - | 8 - 9 | 951 | 101 | |
| AK134849.1 | - | 2 - 3 | 1470 | 101 | ||
| AK148377.1 | - | 6 - 7 | 2257 | 101 | ||
| AK161259.1 | - | 4 - 5 | 1401 | 101 | ||
| BC015080.1 | - | 8 - 9 | 1214 | 3995512 | 101 | |
| CN538691.1 | - | 3 - 4 | 391 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk2
Genomic Map