| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300005486.mm.1 |
NM_001099779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005486.mm.1 |
NM_001099779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005486.mm.1 |
NM_013631.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005486.mm.1 |
NM_013631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005486.mm.1 |
TC0300000747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005487.mm.1 |
NM_001099779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005487.mm.1 |
NM_001099779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005487.mm.1 |
NM_013631.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005487.mm.1 |
NM_013631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005487.mm.1 |
TC0300000747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300005485.mm.1 |
NM_001099779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005485.mm.1 |
NM_001099779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005485.mm.1 |
NM_013631.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005485.mm.1 |
NM_013631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300005485.mm.1 |
TC0300000747.mm.1 |
MTA TC |