Assay Details
| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010251.2 | NP_034381.1 | 2 - 3 | 703 | 94 | |
| GenBank mRNA | AK013727.1 | - | 2 - 3 | 704 | 94 | |
| AK043806.1 | - | 2 - 3 | 321 | 94 | ||
| AK137354.1 | - | 2 - 3 | 319 | 94 | ||
| AK141571.1 | - | 2 - 3 | 637 | 94 | ||
| AK159299.1 | - | 2 - 3 | 664 | 95 | ||
| AK162893.1 | - | 2 - 3 | 318 | 94 | ||
| BC094603.1 | - | 2 - 3 | 331 | 6828002 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabra4
Genomic Map