Assay Details
| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010251.2 | NP_034381.1 | 4 - 5 | 995 | 123 | |
| GenBank mRNA | AK013727.1 | - | 4 - 5 | 996 | 123 | |
| AK043806.1 | - | 4 - 5 | 613 | 123 | ||
| AK139970.1 | - | 4 - 5 | 747 | 123 | ||
| AK141571.1 | - | 4 - 5 | 929 | 123 | ||
| AK159299.1 | - | 4 - 5 | 956 | 123 | ||
| AK162893.1 | - | 4 - 5 | 610 | 123 | ||
| BC094603.1 | - | 4 - 5 | 623 | 6828002 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabra4
Genomic Map