Assay Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040382.1 | 1 - 2 | 49 | 75 | ||
| GenBank mRNA | CJ047138.1 | - | 1 - 2 | 50 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm46os
Genomic Map