Assay Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166531.1 | NP_001160003.1 | 9 - 10 | 1166 | 85 | |
| NM_001166532.1 | NP_001160004.1 | 8 - 9 | 1260 | 85 | ||
| NM_019460.2 | NP_062333.1 | 8 - 9 | 1117 | 85 | ||
| GenBank mRNA | AB032165.1 | - | 7 - 8 | 962 | 85 | |
| AK142330.1 | - | 8 - 9 | 1144 | 85 | ||
| AK154826.1 | - | 8 - 9 | 1262 | 85 | ||
| AK160573.1 | - | 9 - 10 | 1166 | 85 | ||
| BC018217.1 | - | 8 - 9 | 1128 | 85 | ||
| BC036972.1 | - | 8 - 9 | 1187 | 85 | ||
| BC066188.1 | - | 8 - 9 | 1070 | 30476111 | 85 | |
More Information
Set Membership: |
|

Sfmbt1
Genomic Map