Assay Details
| Gene Symbol: Sepsecs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172490.3 | NP_766078.1 | 5 - 6 | 707 | 62 | |
| GenBank mRNA | AK033654.1 | - | 5 - 6 | 708 | 62 | |
| AK049338.1 | - | 714 | 62 | |||
| AK153792.1 | - | 647 | 62 | |||
| BC062133.1 | - | 5 - 6 | 697 | 6516578 | 62 | |
More Information
Set Membership: |
|

Sepsecs
Genomic Map