Assay Details
| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172469.3 | NP_766057.1 | 2 - 3 | 1520 | 86 | |
| GenBank mRNA | AF448440.1 | - | 2 - 3 | 1213 | 86 | |
| AK077599.1 | - | 2 - 3 | 1521 | 86 | ||
| BC075706.1 | - | 2 - 3 | 1545 | 2648653 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clic6
Genomic Map