Assay Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013587.2 | NP_038615.2 | 2 - 3 | 367 | 67 | |
| GenBank mRNA | AJ639614.1 | - | 2 - 3 | 374 | 67 | |
| AK089250.1 | - | 2 - 3 | 370 | 67 | ||
| AK145710.1 | - | 2 - 3 | 410 | 67 | ||
| AK150451.1 | - | 2 - 3 | 381 | 67 | ||
| AK152485.1 | - | 2 - 3 | 381 | 67 | ||
| AK153323.1 | - | 2 - 3 | 381 | 67 | ||
| AK166616.1 | - | 2 - 3 | 370 | 67 | ||
| AK166746.1 | - | 2 - 3 | 381 | 67 | ||
| AK166819.1 | - | 2 - 3 | 385 | 67 | ||
| AK168421.1 | - | 2 - 3 | 370 | 67 | ||
| AK169132.1 | - | 2 - 3 | 375 | 67 | ||
| AK171471.1 | - | 2 - 3 | 370 | 67 | ||
| BC032170.1 | - | 2 - 3 | 370 | 67 | ||
| BC046641.1 | - | 2 - 3 | 359 | 67 | ||
| BC057979.1 | - | 2 - 3 | 338 | 67 | ||
| BC059887.1 | - | 2 - 3 | 184 | 67 | ||
| BC094324.1 | - | 2 - 3 | 367 | 6402163 | 67 | |
| D00622.1 | - | 2 - 3 | 360 | 67 | ||
| S67967.1 | - | 2 - 3 | 393 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrpap1
Genomic Map