| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500020508.mm.1 |
NM_026660.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020508.mm.1 |
NM_026660.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020508.mm.1 |
TC0500002239.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020517.mm.1 |
NM_026660.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020517.mm.1 |
NM_026660.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020517.mm.1 |
TC0500002239.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020521.mm.1 |
TC0500002239.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020511.mm.1 |
NM_026660.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020511.mm.1 |
NM_026660.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020511.mm.1 |
TC0500002239.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500020516.mm.1 |
NM_026660.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020516.mm.1 |
NM_026660.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500020516.mm.1 |
TC0500002239.mm.1 |
MTA TC |