Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 11 - 12 | 1535 | 74 | |
| GenBank mRNA | AF099017.1 | - | 11 - 12 | 1495 | 74 | |
| AF224724.1 | - | 11 - 12 | 1495 | 74 | ||
| AK007987.1 | - | 11 - 12 | 1533 | 74 | ||
| AK075718.1 | - | 11 - 12 | 1444 | 74 | ||
| AK144554.1 | - | 11 - 12 | 1506 | 74 | ||
| AK149522.1 | - | 11 - 12 | 1499 | 74 | ||
| AK165433.1 | - | 9 - 10 | 1884 | 74 | ||
| AK169989.1 | - | 11 - 12 | 1531 | 74 | ||
| BC019376.1 | - | 11 - 12 | 1500 | 5134851 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map