Assay Details
| Gene Symbol: Hgfac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019447.2 | NP_062320.2 | 12 - 13 | 1679 | 124 | |
| GenBank mRNA | AF099017.1 | - | 12 - 13 | 1639 | 124 | |
| AF224724.1 | - | 12 - 13 | 1639 | 124 | ||
| AK007987.1 | - | 12 - 13 | 1677 | 124 | ||
| AK075718.1 | - | 12 - 13 | 1589 | 124 | ||
| AK144554.1 | - | 12 - 13 | 1650 | 124 | ||
| AK149522.1 | - | 12 - 13 | 1643 | 124 | ||
| AK165433.1 | - | 10 - 11 | 2028 | 124 | ||
| AK169989.1 | - | 12 - 13 | 1675 | 124 | ||
| BC019376.1 | - | 12 - 13 | 1644 | 5134851 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgfac
Genomic Map