Assay Details
| Gene Symbol: Nisch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022656.2 | NP_073147.2 | 8 - 9 | 1084 | 73 | |
| GenBank mRNA | AF315344.1 | - | 4 - 5 | 519 | 73 | |
| AK004628.1 | - | 8 - 9 | 921 | 73 | ||
| AK036043.1 | - | 8 - 9 | 939 | 73 | ||
| AK049791.1 | - | 8 - 9 | 939 | 73 | ||
| AK086880.1 | - | 8 - 9 | 939 | 73 | ||
| AK122412.1 | - | 8 - 9 | 1059 | 73 | ||
| AK160990.1 | - | 8 - 9 | 958 | 73 | ||
| BC003270.1 | - | 8 - 9 | 946 | 73 | ||
| BC066838.1 | - | 8 - 9 | 942 | 73 | ||
| BC089329.1 | - | 3 - 4 | 303 | 73 | ||
| BC108364.1 | - | 3 - 4 | 279 | 73 | ||
| BC144980.1 | - | 4 - 5 | 413 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nisch
Genomic Map