Assay Details
| Gene Symbol: Nisch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022656.2 | NP_073147.2 | 9 - 10 | 1158 | 59 | |
| GenBank mRNA | AF315344.1 | - | 5 - 6 | 593 | 59 | |
| AK004628.1 | - | 9 - 10 | 995 | 59 | ||
| AK036043.1 | - | 9 - 10 | 1013 | 59 | ||
| AK049791.1 | - | 9 - 10 | 1013 | 59 | ||
| AK122412.1 | - | 9 - 10 | 1133 | 59 | ||
| AK160990.1 | - | 9 - 10 | 1032 | 59 | ||
| BC003270.1 | - | 9 - 10 | 1020 | 59 | ||
| BC066838.1 | - | 9 - 10 | 1016 | 59 | ||
| BC089329.1 | - | 4 - 5 | 377 | 59 | ||
| BC108364.1 | - | 4 - 5 | 353 | 59 | ||
| BC144980.1 | - | 5 - 6 | 487 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nisch
Genomic Map