Assay Details
| Gene Symbol: Mtcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294277.1 | NP_001281206.1 | 2 - 3 | 244 | 70 | |
| NM_010839.6 | NP_034969.1 | 1 - 2 | 568 | 70 | ||
| GenBank mRNA | BC138811.1 | - | 1 - 2 | 170 | 8861833 | 70 |
| BC138812.1 | - | 1 - 2 | 170 | 8861834 | 70 | |
| DV655269.1 | - | 1 - 2 | 550 | 70 | ||
| Z35294.1 | - | 1 - 2 | 528 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138811.1, BC138812.1, NM_001294277.1, NM_010839.6, Z35294.1 transcripts.
Set Membership: |
|

Mtcp1
Genomic Map