Assay Details
| Gene Symbol: Adgra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054044.2 | NP_473385.2 | 9 - 10 | 1400 | 68 | |
| GenBank mRNA | AF378759.1 | - | 9 - 10 | 1400 | 68 | |
| BC138451.1 | - | 9 - 10 | 1390 | 8861471 | 68 | |
| BC138452.1 | - | 9 - 10 | 1390 | 8861472 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgra3
Genomic Map