Assay Details
| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160403.1 | NP_001153875.1 | 4 - 5 | 1193 | 96 | |
| GenBank mRNA | AK081272.1 | - | 4 - 5 | 1260 | 96 | |
| BC119580.2 | - | 4 - 5 | 1193 | 40058380 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rapl1
Genomic Map