Assay Details
| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160403.1 | NP_001153875.1 | 9 - 10 | 1847 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rapl1
Genomic Map