Assay Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356878.1 | - | 5 - 6 | 717 | 100 | |
| AF356879.1 | - | 4 - 5 | 576 | 100 | ||
| AF375479.1 | - | 3 - 4 | 528 | 100 | ||
| AK002247.1 | - | 5 - 6 | 716 | 100 | ||
| AK143782.1 | - | 5 - 6 | 719 | 100 | ||
| AY169234.1 | - | 5 - 6 | 716 | 100 | ||
| BC010795.1 | - | 3 - 4 | 937 | 4221711 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acy3
Genomic Map