Assay Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356878.1 | - | 7 - 8 | 932 | 68 | |
| AF356879.1 | - | 6 - 7 | 791 | 68 | ||
| AF375479.1 | - | 5 - 6 | 743 | 68 | ||
| AK002247.1 | - | 7 - 8 | 931 | 68 | ||
| AK143782.1 | - | 7 - 8 | 934 | 68 | ||
| AY169234.1 | - | 7 - 8 | 931 | 68 | ||
| BC010795.1 | - | 5 - 6 | 1152 | 4221711 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118411 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103600 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103600 | - | - |
| Whole Mouse Genome G4122A | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AY169234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | BC010795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AF356878.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AF356879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232399 | AK002247.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140237 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100088 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14334336 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14334322 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14334334 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14334324 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14182134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14334327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14334336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14317851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14453632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14317864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14453638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14317860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14375590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14182128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14284799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14143321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14334335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460272 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448539_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460263 | - | - |
| Mouse Genome 430 2.0 Array | 1448539_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448539_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71670_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71670_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000249.mm.1 | TC1900000022.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448539_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448539_a_at | - | - |
| Murine Genome U74v2 Set | 95725_at | - | - |
| Murine Genome U74v2 Set | 95725_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448539_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448539_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448539_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448539_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6110369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6110369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240165 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240165 | - | - |
| mouse-6 v1.1 (Illumina) | scl52796.6.1_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52796.6.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240165 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240165 | - | - |

Acy3
Genomic Map