Assay Details
| Gene Symbol: Cabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160252.1 | NP_001153724.1 | 3 - 4 | 262 | 78 | |
| NM_001160253.1 | NP_001153725.1 | 3 - 4 | 544 | 78 | ||
| NM_013878.2 | NP_038906.2 | 4 - 5 | 421 | 78 | ||
| GenBank mRNA | AF169156.1 | - | 3 - 4 | 290 | 78 | |
| AF169157.1 | - | 4 - 5 | 449 | 78 | ||
| AK007488.1 | - | 3 - 4 | 545 | 78 | ||
| BC107029.2 | - | 4 - 5 | 421 | 40050356 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cabp2
Genomic Map