Assay Details
| Gene Symbol: Dlgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145965.1 | NP_001139437.1 | 3 - 4 | 354 | 59 | |
| NM_172910.3 | NP_766498.2 | 4 - 5 | 453 | 59 | ||
| GenBank mRNA | AK032564.1 | - | 4 - 5 | 454 | 59 | |
| AK082945.1 | - | 2 - 3 | 226 | 59 | ||
| BB622956.1 | - | 454 | 59 | |||
| BC118969.2 | - | 3 - 4 | 140 | 40097764 | 59 | |
More Information
Set Membership: |
|

Dlgap2
Genomic Map