Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172993.3 | NP_766581.2 | 5 - 6 | 582 | 76 | |
| GenBank mRNA | AK084205.1 | - | 5 - 6 | 529 | 76 | |
| AK146353.1 | - | 5 - 6 | 546 | 76 | ||
| AK149921.1 | - | 5 - 6 | 529 | 76 | ||
| AK154918.1 | - | 5 - 6 | 529 | 76 | ||
| AK173267.1 | - | 5 - 6 | 557 | 76 | ||
| BC021520.1 | - | 3 - 4 | 392 | 76 | ||
| BC066136.1 | - | 5 - 6 | 505 | 76 | ||
| BC089160.1 | - | 5 - 6 | 504 | 5390706 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp512
Genomic Map