Assay Details
| Gene Symbol: Ccr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007718.3 | NP_031744.3 | 1 | 836 | 72 | |
| GenBank mRNA | BC127143.1 | - | 1 | 836 | 40051964 | 72 |
| BC128466.1 | - | 1 | 836 | 40051965 | 72 | |
| CF551376.1 | - | 1 | 108 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccr1l1
Genomic Map