Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289798.1 | NP_001276727.1 | 2 - 3 | 200 | 103 | |
| NM_001289799.1 | NP_001276728.1 | 2 - 3 | 196 | 103 | ||
| NM_145558.2 | NP_663533.1 | 2 - 3 | 153 | 103 | ||
| GenBank mRNA | AK033462.1 | - | 2 - 3 | 135 | 103 | |
| AK083164.1 | - | 2 - 3 | 161 | 103 | ||
| AK083767.1 | - | 2 - 3 | 116 | 103 | ||
| AK150889.1 | - | 2 - 3 | 115 | 103 | ||
| AK168594.1 | - | 2 - 3 | 117 | 103 | ||
| AK169637.1 | - | 2 - 3 | 129 | 103 | ||
| AK171803.1 | - | 2 - 3 | 118 | 103 | ||
| BC005585.1 | - | 1 - 2 | 84 | 3158015 | 103 | |
| BY337615.1 | - | 2 - 3 | 154 | 103 | ||
| BY778233.1 | - | 2 - 3 | 130 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK033462.1,AK083164.1,AK083767.1,AK150889.1,AK169637.1,BC005585.1,NM_001289798.1,NM_001289799.1,NM_145558.2 transcripts.
This assay is in the 5'UTR region of the AK033462.1,AK083164.1,AK083767.1,AK150889.1,AK169637.1,BC005585.1,NM_001289798.1,NM_001289799.1,NM_145558.2 transcripts.
Set Membership: |
|

Hadhb
Genomic Map